Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRPML3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564006
Description
Immunogen sequence: KGYMDRMDDT YAVYTQSDVY DQLIFAVNQY LQLYNVSVGN HAY Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.
Expressed in the cochlea; particularly in the inner and outer hair cells. Defects in Mcoln3 are the cause of the varitin-waddler (Va) phenotype. Classical Va mice exhibit early-onset hearing loss, vestibular defects, pigmentation abnormalities and perinatal lethality. The phenotype varitin-waddler Jackcon (Va-J), which arose in a cross segregating for Va, is similar but less severe. Belongs to the transient receptor family, polycystin subfamily.
Specifications
| TRPML3 | |
| Polyclonal | |
| Unconjugated | |
| MCOLN3 | |
| 6720490O21Rik; FLJ11006; FLJ36629; MCLN3; Mcoln3; Mcoln3 mucolipin 3; MGC124245; MGC124246; MGC71509; mucolipin 3; mucolipin-3; Transient receptor potential channel mucolipin 3; TRPML3; TRP-ML3; Va; varitint-waddler | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55283 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TDD5 | |
| MCOLN3 | |
| Recombinant protein corresponding to Human Mucolipin 3. Recombinant protein control fragment (Product #RP-103436). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction