Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRPML3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109339
Description
Positive Control: 293 cell, F9.
Expressed in the cochlea; particularly in the inner and outer hair cells. Defects in Mcoln3 are the cause of the varitin-waddler (Va) phenotype. Classical Va mice exhibit early-onset hearing loss, vestibular defects, pigmentation abnormalities and perinatal lethality. The phenotype varitin-waddler Jackcon (Va-J), which arose in a cross segregating for Va, is similar but less severe. Belongs to the transient receptor family, polycystin subfamily.
Specifications
TRPML3 | |
Polyclonal | |
Unconjugated | |
MCOLN3 | |
6720490O21Rik; FLJ11006; FLJ36629; MCLN3; Mcoln3; Mcoln3 mucolipin 3; MGC124245; MGC124246; MGC71509; mucolipin 3; mucolipin-3; Transient receptor potential channel mucolipin 3; TRPML3; TRP-ML3; Va; varitint-waddler | |
Rabbit | |
Affinity chromatography | |
RUO | |
171166, 55283 | |
-20°C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8R4F0, Q8TDD5 | |
MCOLN3 | |
Synthetic peptide within Human TRPML3 aa 504-553. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction