Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPS1 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204120UV
Description
TRPS1 Polyclonal antibody specifically detects TRPS1 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
TRPS1 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
trichorhinophalangeal syndrome I, tricho-rhino-phalangeal syndrome type I protein | |
The immunogen recognized by this antibody maps to a region between residue 1000 and 1050 of human Tricho-Rhino-Phalangeal Syndrome Type I using the numbering given in entry AAG21134.1 (GeneID 7227). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
7227 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction