Learn More
Invitrogen™ TRPS1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total TRPS1 protein.
Specifications
Specifications
| Antigen | TRPS1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TRPS1 |
| Gene Accession No. | Q925H1, Q9UHF7 |
| Gene Alias | AI115454; AI447310; D15Ertd586e; GC79; LGCR; transcriptional repressor GATA binding 1; trichorhinophalangeal syndrome I; trichorhinophalangeal syndrome I (human); trichorhinophalangeal syndrome I homolog; tricho-rhino-phalangeal syndrome type I protein; TRPS1; Zinc finger protein GC79; zinc finger transcription factor Trps1 |
| Gene Symbols | TRPS1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.