Learn More
Invitrogen™ TRPS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596784
Description
Immunogen sequence: PPLRNVASEG EGQILEPIGT ESKVSGKNKE FSADQMSENT DQSDAAELNH KEEHSLHVQD PSSSSKKDLK SAVLSEKAGF NYESPSKGGN FPSFPHDEVT DRNMLAFSSP AAGGVCEPLK SPQRAEADDP QDMACTPSGD SLETKEDQKM SPKATEETGQ AQSGQANCQG LSPVSVASKN PQVPSDGGVR LNKSKTDLLV NDNPDPAPLS PELQDFKCNI CGYGYYGNDP TDLIKHFRKY H; Positive Samples: Mouse kidney, Mouse spleen; Cellular Location: Nucleus.
This gene encodes a transcription factor that represses GATA-regulated genes and binds to a dynein light chain protein. Binding of the encoded protein to the dynein light chain protein affects binding to GATA consensus sequences and suppresses its transcriptional activity. Defects in this gene are a cause of tricho-rhino-phalangeal syndrome (TRPS) types I-III. [provided by RefSeq].
Specifications
TRPS1 | |
Polyclonal | |
Unconjugated | |
TRPS1 | |
AI115454; AI447310; D15Ertd586e; GC79; LGCR; transcriptional repressor GATA binding 1; trichorhinophalangeal syndrome I; trichorhinophalangeal syndrome I (human); trichorhinophalangeal syndrome I homolog; tricho-rhino-phalangeal syndrome type I protein; TRPS1; Zinc finger protein GC79; zinc finger transcription factor Trps1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
7227, 83925 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q925H1, Q9UHF7 | |
TRPS1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 20-260 of human TRPS1 (NP_0548312). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.