Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106538
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106538 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106538 Supplier Invitrogen™ Supplier No. PA5106538
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TRPS1.

This gene encodes a transcription factor that represses GATA-regulated genes and binds to a dynein light chain protein. Binding of the encoded protein to the dynein light chain protein affects binding to GATA consensus sequences and suppresses its transcriptional activity. Defects in this gene are a cause of tricho-rhino-phalangeal syndrome (TRPS) types I-III. [provided by RefSeq].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TRPS1
Gene Accession No. Q925H1, Q9UHF7
Gene Alias AI115454; AI447310; D15Ertd586e; GC79; LGCR; transcriptional repressor GATA binding 1; trichorhinophalangeal syndrome I; trichorhinophalangeal syndrome I (human); trichorhinophalangeal syndrome I homolog; tricho-rhino-phalangeal syndrome type I protein; TRPS1; Zinc finger protein GC79; zinc finger transcription factor Trps1
Gene Symbols TRPS1
Host Species Rabbit
Immunogen A synthesized peptide derived from human TRPS1(Accession Q9UHF7), corresponding to amino acid residues K126-G176.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7227, 83925
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.