Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | TRPT1 |
|---|---|
| Concentration | 0.15 mg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0299
![]()
|
Proteintech Group Inc
155411AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
TRPT1 catalyzes the last step of tRNA splicing, the transfer of the splice junction 2′-phosphate from ligated tRNA to NAD to produce ADP-ribose 1′′-2′′ cyclic phosphate. It has four isoforms with MW 22-24 and 27-29 kDa.Specifications
| TRPT1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q86TN4, Q8K3A2 | |
| 107328, 293704, 83707 | |
| TRPT1 Fusion Protein Ag7897 | |
| Primary | |
| -20°C | |
| TRPT1 |
| 0.15 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| tRNA 2 phosphotransferase 1, tRNA phosphotransferase 1, TRPT1 | |
| Trpt1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title