Learn More
TRPV1 Polyclonal Antibody, Invitrogen™
Guinea Pig Polyclonal Antibody
Supplier: Thermo Scientific PA5111831
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)EDAEVFK DSMVPGEK Homology: Mouse - 14/15 amino acid residues identical; human - 10/15 amino acid residues identical.
TRPV1 (Transient Receptor Potential Cation Channel Subfamily V Member 1) is a receptor for capsaicin, the main pungent ingredient in hot chili peppers, and elicits a sensation of burning pain by selectively activating sensory neurons. TRPV1 is a non-selective cation channel that is structurally related to members of the TRP family of ion channels such as TRPC3 and TRPC6. TRPV1 is permeable to calcium, magnesium, and monovalent cations of sodium and potassium. Structurally, TRPV1 contains 6 trans-membrane domains, a P-loop membrane domain and 3 ankyrin repeat domains. TRPV1 is also activated by increases in temperature in the noxious range, suggesting that it functions as a transducer of painful thermal stimuli in vivo. TRPV1 seems to mediate proton influx and may be involved in intracellular acidosis in nociceptive neurons. The expression of TRPV1 appears to be limited to neurons within the sensory ganglia. Four transcript variants encoding TRPV1 with variations in th 5′ UTR sequence have been described. Diseases associated with TRPV1 include Irritable Bowel Syndrome (IBS) and Bladder Disease.
Specifications
TRPV1 | |
Polyclonal | |
Unconjugated | |
TRPV1 | |
Capsaicin receptor; DKFZp434K0220; Osm-9-like TRP channel 1; OTRPC1; Transient receptor potential cation channel subfamily V member 1; TRPV1; TRPV1_SON; TRPV1alpha; TRPV1beta; Vanilloid receptor; Vanilloid receptor 1; Vanilloid receptor type 1-like; VR.5′sv; VR1; VR-1; Vr1l; Vr1l1 | |
Guinea Pig | |
Antigen affinity chromatography | |
RUO | |
83810 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
0.85 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
O35433 | |
TRPV1 | |
Peptide corresponding to amino acid residues 824-838 of rat TRPV1 (Intracellular, C-terminus) | |
50 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.