Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPV2 Mouse anti-Mouse, Rat, Unconjugated, Clone: 17A11, MilliporeSigma™

Catalog No. MABN242525 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2425-25 25 μg
MABN2425-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN2425-25 Supplier MilliporeSigma Supplier No. MABN242525UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-TRPV2, clone 17A11, Cat. No. MABN2425, is a highly specific mouse monoclonal antibody that targets TRPV2 and has been tested for use in Immunocytochemistry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPV2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 17A11
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Transient receptor potential cation channel subfamily V member 2;Osm-9-like TRP channel 2;OTRPC2;Stretch-activated channel 2B;Vanilloid receptor-like protein 1;VRL-1
Gene Symbols Trpv2;Sac2b;Vrl1
Host Species Mouse
Immunogen 1D4-tagged full length rat Transient receptor potential cation channel subfamily V member 2 (TRPV2).
Molecular Weight of Antigen 86.71 kDa calculated.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_058903.2
Test Specificity Clone 17A11 detects Transient receptor potential cation channel subfamily V member 2 (TRPV2) in mouse and rat cells.
Target Species Mouse, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.