Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPV2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533091
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533091 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533091 Supplier Invitrogen™ Supplier No. PA533091
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Horse, Pig (100%); Gibbon, Bovine, Hamster, Panda, Rabbit (93%); Mouse, Dog (87%); Elephant (80%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

VLR-1 is a calcium-permeable, non-selective cation channel with an outward rectification, that seems to be regulated, at least in part, by growth factors, like IGF1, PDGF and morphogenetic neuropeptide/head activator. VRL-1 does not respond to capsaicin, acid, or moderate heat, but instead is activated by high temperatures, with a threshold of approximately 52 degrees C. Within sensory ganglia, VRL-1 is most prominently expressed by a subset of medium- to large-diameter neurons, making it a candidate receptor for transducing high-threshold heat responses in this class of cells. VRL-1 transcripts are not restricted to the sensory nervous system, indicating that this channel may be activated by stimuli other than heat. VRL-1 is one of the six members of the vanilloid-like TRP-channel family which is now known as TRPV family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPV2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Trpv2
Gene Accession No. Q9Y5S1
Gene Alias GRC; growth factor regulated channel; growth factor-regulated calcium channel; Osm-9-like TRP channel 2; OTRPC2; RP23-234K24.7; Sac2b; Stretch-activated channel 2B; transient receptor potential cation channel subfamily V member 2; transient receptor potential cation channel, subfamily V, member 2; Trpv2; Vanilloid receptor-like protein 1; VRL; Vrl1; VRL-1
Gene Symbols Trpv2
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from N-terminus of human TRPV2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100063195, 100523638, 51393
Target Species Human, Monkey, Pig, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.