Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPV4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
$206.50 - $548.00
Specifications
Antigen | TRPV4 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1: 1000 - 1: 2000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018021
![]() |
Novus Biologicals
NB110749600.025ML |
0.025 mL |
Each for $206.50
|
|
|||||
NB11074960
![]() |
Novus Biologicals
NB11074960 |
0.1 mL |
Each for $548.00
|
|
|||||
Description
TRPV4 Polyclonal specifically detects TRPV4 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TRPV4 | |
Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1: 1000 - 1: 2000 | |
Polyclonal | |
Antisera | |
RUO | |
Q9EPK8 | |
59341 | |
IgG | |
Unpurified | |
98 kDa |
LYOPH | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Monkey, Mouse, Rat | |
CMT2Cosmosensitive transient receptor potential channel 4, HMSN2C, Osm-9-like TRP channel 4, OTRPC4SSQTL1, transient receptor potential cation channel subfamily V member 4, transient receptor potential cation channel, subfamily V, member 4, Transient receptor potential protein 12, TRP12OSM9-like transient receptor potential channel 4, TrpV4, Vanilloid receptor-like channel 2, Vanilloid receptor-like protein 2, vanilloid receptor-related osmotically activated channel, Vanilloid receptor-related osmotically-activated channel, VRL2, VRL-2, VR-OAC, VROACSPSMA | |
TRPV4 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title