Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPV4 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241262PCP
Description
TRPV4 Polyclonal antibody specifically detects TRPV4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| TRPV4 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| CMT2Cosmosensitive transient receptor potential channel 4, HMSN2C, Osm-9-like TRP channel 4, OTRPC4SSQTL1, transient receptor potential cation channel subfamily V member 4, transient receptor potential cation channel, subfamily V, member 4, Transient receptor potential protein 12, TRP12OSM9-like transient receptor potential channel 4, TrpV4, Vanilloid receptor-like channel 2, Vanilloid receptor-like protein 2, vanilloid receptor-related osmotically activated channel, Vanilloid receptor-related osmotically-activated channel, VRL2, VRL-2, VR-OAC, VROACSPSMA | |
| Antibody was raised against an 18 amino acid peptide near the center of human TRPV4. The immunogen is located within amino acids 380 - 430 of TRPV4. Amino Acid Squence: RREVTDEDTRHLSRKFKD | |
| 0.1 mL | |
| Neuroscience | |
| 59341 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction