Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPV5 Antibody (SR2284), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322511100UL
Description
TRPV5 Monoclonal antibody specifically detects TRPV5 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
TRPV5 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
SR2284 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
Calcium transport protein 2, calcium transporter 2, ECaC, ECaC1, ECAC1CaT2, Epithelial calcium channel 1CAT2, Osm-9-like TRP channel 3, OTRPC3, transient receptor potential cation channel subfamily V member 5, transient receptor potential cation channel, subfamily V, member 5, TrpV5 | |
A synthesized peptide derived from human TRPV5 (Uniprot #: Q9NQA5) | |
100 μg | |
Cancer, Endocrinology, Signal Transduction | |
56302 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction