Learn More
Invitrogen™ TRPV5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595269
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Pancreas Tissue, Rat Lung Tissue, Rat Intestine Tissue, SW620 whole cell, COLO320 whole cell, 293T whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This gene is a member of the transient receptor family and the TrpV subfamily. The calcium-selective channel encoded by this gene has 6 transmembrane-spanning domains, multiple potential phosphorylation sites, an N-linked glycosylation site, and 5 ANK repeats. This protein forms homotetramers or heterotetramers and is activated by a low internal calcium level.
Specifications
TRPV5 | |
Polyclonal | |
Unconjugated | |
TRPV5 | |
Calciu; Calcium transport protein 2; calcium transporter 2; CaT2; D630033B11; ECAC; Ecac1; epithelial calcium channel; epithelial calcium channel 1; Osm-9-like TRP channel 3; OTRPC3; Transient receptor potential cation channel subfamily V member 5; transient receptor potential cation channel, subfamily V, member 5; Trpv5 | |
Rabbit | |
Affinity chromatography | |
RUO | |
116469, 56302 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
Q9JIP0, Q9NQA5 | |
TRPV5 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human TRPV5 (580-610aa DTHWRVAQERDELWRAQVVATTVMLERKLPR). | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.