Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPV6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144631
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144631 Supplier Invitrogen™ Supplier No. PA5144631
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: SW480 cell lysate, SK-Br-3 cell lysate, HepG2, HT-29, rat testis tissue, human kidney tissue, mouse colon tissue, human placenta tissue. Predicted band size: 87 kDa Subcellular Location: Cell membrane, Membrane.

Calcium-permeable channels, such as TRPV6, participate in neurotransmission, muscle contraction, and exocytosis by providing calcium as an intracellular second messenger. Depending on the tissue, transcellular calcium transport may be regulated by vitamin D, parathyroid hormone (PTH), or calcitonin (CALCA). Calcium selective cation channel probably involved in Ca(2+) uptake in various tissues, including Ca(2+) reabsorption in intestine. The channel is activated by low internal calcium level, probably including intracellular calcium store depletion, and the current exhibits an inward rectification. Inactivation includes both, a rapid Ca(2+)-dependent and a slower Ca(2+)-calmodulin-dependent mechanism, the latter may be regulated by phosphorylation. In vitro, is slowly inhibited by Mg(2+) in a voltage-independent manner. Heteromeric assembly with TRPV5 seems to modify channel properties. TRPV5-TRPV6 heteromultimeric concatemers exhibit voltage-dependent gating. Interacts with TRPV5. Interacts with S100A10 and probably with the ANAX2-S100A10 heterotetramer. The interaction with S100A10 is required for the trafficking to the plasma membrane. Interacts with BSPRY. Interacts with calmodulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPV6
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TRPV6
Gene Accession No. Q91WD2, Q9H1D0, Q9R186
Gene Alias ABP/ZF; Alu-binding protein with zinc finger domain; Cac; Calcium transport protein 1; calcium transporter-like protein; CAT; CaT1; CATL; CaT-L; CaT-like; ecac; ECaC; CaT; ECAC2; epithelial apical membrane calcium transporter/channel CaT1; epithelial calcium channel; epithelial calcium channel 2; HSA277909; ion channel; LP6728; matt-und-schlapp; mus; osmosensitive transient receptor potential channel 3; Otrpc3; transient receptor potential cation channel subfamily V member 6; transient receptor potential cation channel, subfamily 5, member 6; transient receptor potential cation channel, subfamily V, member 6; transient receptor potential vanilloid 5-6; trpv5-6; TRPV6; trpv6 protein; ZFAB
Gene Symbols TRPV6
Host Species Rabbit
Immunogen Synthetic peptide corresponding to C terminal of Human TRPV6.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114246, 55503, 64177
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.