Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPV6 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502209530
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9530 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9530 Supplier Fabgennix Inc Supplier No. TRPV6601AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRPV6 Polyclonal antibody specifically detects TRPV6 in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen TRPV6
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene TRPV6
Gene Accession No. Q91WD2, Q9H1D0, Q9R186
Gene Alias ABP/ZF; Alu-binding protein with zinc finger domain; Cac; Calcium transport protein 1; calcium transporter-like protein; CAT; CaT1; CATL; CaT-L; CaT-like; ecac; ECaC; CaT; ECAC2; epithelial apical membrane calcium transporter/channel CaT1; epithelial calcium channel; epithelial calcium channel 2; HSA277909; ion channel; LP6728; matt-und-schlapp; mus; osmosensitive transient receptor potential channel 3; Otrpc3; transient receptor potential cation channel subfamily V member 6; transient receptor potential cation channel, subfamily 5, member 6; transient receptor potential cation channel, subfamily V, member 6; transient receptor potential vanilloid 5-6; trpv5-6; TRPV6; trpv6 protein; ZFAB
Host Species Rabbit
Immunogen Synthetic peptide taken from the region within amino acids 700-732 of TRPV6 protein. The peptide was covalently modified to acheive desired antigenicity. Modified peptide was covalently coupled to a carrier protein by heterobifunctional cross linker for immunogen preparation.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114246, 55503, 64177
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.