Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRRAP Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10058810AF594
Description
TRRAP Polyclonal antibody specifically detects TRRAP in Human samples. It is validated for Western BlotSpecifications
TRRAP | |
Polyclonal | |
Western Blot | |
350/400 kDa PCAF-associated factor, FLJ10671, PAF350/400, PAF400STAF40, Tra1, Tra1 homolog, transformation/transcription domain-associated protein, TR-AP | |
The immunogen recognized by this antibody maps to a region between residue 3275 and 3325 of human transformation/transcription domain-associated protein using the numbering given in entry AAD04629.1 (GeneID 8295). | |
0.1 mL | |
Cell Cycle and Replication, Protein Kinase | |
8295 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction