Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRRAP Antibody, Novus Biologicals™
SDP

Catalog No. NB10058811 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10058811 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10058811 Supplier Novus Biologicals Supplier No. NB10058811
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRRAP Polyclonal specifically detects TRRAP in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen TRRAP
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 mcg/mg
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. AAD04629.1
Gene Alias 350/400 kDa PCAF-associated factor, FLJ10671, PAF350/400, PAF400STAF40, Tra1, Tra1 homolog, transformation/transcription domain-associated protein, TR-AP
Gene Symbols TRRAP
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 3809 and 3859 of human transformation/transcription domain-associated protein using the numbering given in entry AAD04629.1 (GeneID 8295).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 8295
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.