Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRRAP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | TRRAP |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
TRRAP Polyclonal specifically detects TRRAP in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
TRRAP | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication, Protein Kinase | |
350/400 kDa PCAF-associated factor, FLJ10671, PAF350/400, PAF400STAF40, Tra1, Tra1 homolog, transformation/transcription domain-associated protein, TR-AP | |
TRRAP | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
8295 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TWVQLFPRLWKILSDRQQHALAGEISPFLCSGSHQVQRDCQPSALNCFVEAMSQCVPPIPIRPCVLKYLGKTHNLWFRSTLMLEHQ | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title