Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TRRAP, Mouse, Unlabeled, Clone: 2TRR-1B3,

Catalog No. MABE1008MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABE1008MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1008MI Supplier MilliporeSigma™ Supplier No. MABE1008
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Platelet factor 4 (UniProt P02776; also known as C-X-C motif chemokine 4, Iroplact, Oncostatin-A, PF-4) is encoded by the PF4 (also known as CXCL4, SCYB4) gene (Gene ID 5196) in human. Platelet factor 4 (PF4) is a small CXC chemokine family cytokine released from alpha-granules of activated platelets during platelet aggregation to promote blood coagulation. PF4 binds heparin with high affinity (Kd ∽4-20 nM) via its heparin-binding domain (a.a. 92-98) and neutralizes the anti-thrombin activity of heparin-like molecules. PF4 is chemotactic for neutrophils, fibroblasts and monocytes, and interacts with the chemokine receptor CXCR3 splice variant CXCR3B. In individuals suffering from heparin-induced thrombocytopenia (HIT), PF4-heparin complexes can trigger an immune response and the generation of anti-PF4 antibodies, resulting in the foramtion of macromolecular antibody-heparin-PF4 immune complexes that bind Fc RIIa (CD32a) on platelets and induce platelet activation. Platelet activation in turn results in more PF4 release into circulation, enhanced immune-complex formation, exaggerated platelet activation, and cell clearance. PF4 is synthesized with a signal peptide sequence (a.a. 1-31) that is removed posttranslationally to produce the mature PF4 (a.a. 32-101), additional proteolytic cleavage yields the PF4 short form (a.a. 48-101).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRRAP
Applications Western Blot
Classification Monoclonal
Clone 2TRR-1B3
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Mouse monoclonal IgG2aκ ascites without preservatives.
Gene TRRAP, PAF400
Gene Accession No. Q9Y4A5
Host Species Mouse
Immunogen Ovalbumin-conjugated linear peptide corresponding to a sequence from the TP53-interacting region of human TRRAP.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001231509
Test Specificity The immunogen sequence is present in both spliced isoforms of human TRRAP reported by UniProt (Q9Y4A5).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Ascites
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.