Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRRAP Polyclonal Antibody
TRRAP Polyclonal Antibody
Supplier: Thermo Scientific PA517187
Description
TRRAP Polyclonal Antibody for Western Blot

Specifications
TRRAP | |
Polyclonal | |
Unconjugated | |
TRRAP | |
350/400 kDa PCAF-associated factor; AI481500; OTTHUMP00000197294; OTTHUMP00000197295; PAF350/400; PAF400; STAF40; Tra1; Tra1 homolog; transactivation/transformation-domain associated protein; transformation/transcription domain associated protein; transformation/transcription domain-associated protein; TR-AP; TRRAP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100683, 288471, 8295 | |
-20°C | |
Liquid |
Western Blot | |
125.4 μg/mL | |
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
Q80YV3, Q9Y4A5 | |
TRRAP | |
Synthetic peptide corresponding to the human TRRAP protein. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction