Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRSPAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556914
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556914 Supplier Invitrogen™ Supplier No. PA556914
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLNYATYGKQ PDNSPEYSLF VGDLTPDVDD GMLYEFFVKV YPSCRGGKVV LDQTGVSKGY GFVKFTDELE QK Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 100%.

TRSPAP1 is involved in the early steps of selenocysteine biosynthesis and tRNA(Sec) charging to the later steps resulting in the cotranslational incorporation of selenocysteine into selenoproteins. It stabilizes the SECISBP2, EEFSEC and tRNA(Sec) complex. It may be involved in the methylation of tRNA(Sec) and enhances efficiency of selenoproteins synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRSPAP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRNAU1AP
Gene Accession No. Q9NX07
Gene Alias 1110007F05Rik; AA387999; FLJ20503; PRO1902; RP4-669K10.4; SECP43; tRNA selenocysteine 1 associated protein 1; tRNA selenocysteine 1-associated protein 1; tRNA selenocysteine associated protein; tRNA selenocysteine associated protein (SECP43); tRNA selenocysteine associated protein 1; tRNA selenocysteine-associated protein 1; TRNAU1AP; Trspap1; Unknown (protein for MGC:140234); WI1-2643G14.2
Gene Symbols TRNAU1AP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRSPAP1. Recombinant protein control fragment (Product #RP-94478).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54952
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.