Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRSPAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590483
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590483 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590483 Supplier Invitrogen™ Supplier No. PA590483
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAASLWMGDL EPYMDENFIS RAFATMGETV MSVKIIRNRL TGIPAGYCFV EFADLATAEK CLHKINGKPL PGATPAKRFK LNYATYGKQP DNSPEYSLFV GDLTPDVDDG MLYEFFVKVY PSCRGGKVVL DQTGVSKGYG FVKFTDELEQ KRALTECQGA VGLGSKPVRL SVAIPKASRV KPVEYSQMYS YSYNQYYQQY QNYYAQWGYD QNTGSYSYSY PQYGYTQSTM QTYEEVGDDA LEDPMPQLDV TEANKEFMEQ SEELYDALMD CHWQPLDTVS SEIPAMM; Positive Samples: A-549, Mouse lung; Cellular Location: Cytoplasm, Nucleus.

TRSPAP1 is involved in the early steps of selenocysteine biosynthesis and tRNA(Sec) charging to the later steps resulting in the cotranslational incorporation of selenocysteine into selenoproteins. It stabilizes the SECISBP2, EEFSEC and tRNA(Sec) complex. It may be involved in the methylation of tRNA(Sec) and enhances efficiency of selenoproteins synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRSPAP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TRNAU1AP
Gene Accession No. Q80VC6, Q9NX07
Gene Alias 1110007F05Rik; AA387999; FLJ20503; PRO1902; RP4-669K10.4; SECP43; tRNA selenocysteine 1 associated protein 1; tRNA selenocysteine 1-associated protein 1; tRNA selenocysteine associated protein; tRNA selenocysteine associated protein (SECP43); tRNA selenocysteine associated protein 1; tRNA selenocysteine-associated protein 1; TRNAU1AP; Trspap1; Unknown (protein for MGC:140234); WI1-2643G14.2
Gene Symbols TRNAU1AP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-287 of human TRNAU1AP (NP_0603161).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54952, 71787
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.