Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRSPAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113438
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113438 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113438 Supplier Invitrogen™ Supplier No. PA5113438
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRSPAP1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRSPAP1 is involved in the early steps of selenocysteine biosynthesis and tRNA(Sec) charging to the later steps resulting in the cotranslational incorporation of selenocysteine into selenoproteins. It stabilizes the SECISBP2, EEFSEC and tRNA(Sec) complex. It may be involved in the methylation of tRNA(Sec) and enhances efficiency of selenoproteins synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRSPAP1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TRNAU1AP
Gene Accession No. Q9NX07
Gene Alias 1110007F05Rik; AA387999; FLJ20503; PRO1902; RP4-669K10.4; SECP43; tRNA selenocysteine 1 associated protein 1; tRNA selenocysteine 1-associated protein 1; tRNA selenocysteine associated protein; tRNA selenocysteine associated protein (SECP43); tRNA selenocysteine associated protein 1; tRNA selenocysteine-associated protein 1; TRNAU1AP; Trspap1; Unknown (protein for MGC:140234); WI1-2643G14.2
Gene Symbols TRNAU1AP
Host Species Rabbit
Immunogen Recombinant Human tRNA selenocysteine 1-associated protein 1 protein (33-156AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54952
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.