Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRUB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548450
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548450 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548450 Supplier Invitrogen™ Supplier No. PA548450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRUB1 Polyclonal Antibody for Western Blot, IHC (P)

Pseudouridine is an abundant component of rRNAs and tRNAs and is enzymatically generated by isomerization of uridine by pseudouridine synthase (Zucchini et al., 2003 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRUB1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TRUB1
Gene Accession No. Q8WWH5
Gene Alias 2610009I02Rik; 9030425C13Rik; AI448235; BB118943; C76870; probable tRNA pseudouridine synthase 1; PUS4; TruB pseudouridine (psi) synthase family member 1; TruB pseudouridine (psi) synthase homolog 1; TruB pseudouridine (psi) synthase homolog 1 (E. coli); TruB pseudouridine synthase family member 1; TRUB1
Gene Symbols TRUB1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 152-180 amino acids from the Central region of human TRUB1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 142940
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.