Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRUB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590731
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590731 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590731 Supplier Invitrogen™ Supplier No. PA590731
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AHLTKDYTVR GLLGKATDDF REDGRLVEKT TYDHVTREKL DRILAVIQGS HQKALVMYSN LDLKTQEAYE MAVRGLIRPM NKSPMLITGI RCLYFAPPEF LLEVQCMHET QKELRKLVHE IGLELKTTAV CTQVRRTRDG FFTLDSALLR TQWDLTNIQD AIRAATPQVA AELEKSLSPG LDTKQLPSPG WSWDSQGPSS TLGLERGAGQ; Positive Samples: K-562, HeLa.

Pseudouridine is an abundant component of rRNAs and tRNAs and is enzymatically generated by isomerization of uridine by pseudouridine synthase (Zucchini et al., 2003 [PubMed 12736709]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRUB2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TRUB2
Gene Accession No. O95900
Gene Alias CLONE24922; G430055L02Rik; Mitochondrial mRNA pseudouridine synthase TRUB2; probable tRNA pseudouridine synthase 2; RP11-339B21.1; TruB pseudouridine (psi) synthase family member 2; TruB pseudouridine (psi) synthase homolog 2; TruB pseudouridine (psi) synthase homolog 2 (E. coli); TruB pseudouridine synthase family member 2; Trub2
Gene Symbols TRUB2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 122-331 of human TRUB2 (NP_0564941).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26995
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.