Learn More
Perfex TruCLEAN 2
Catalog No. p-200003936
Click to view available options
Color:
Green
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy
Unique bucket-in-bucket design to simplify standard operating procedure and maximize operator adaptability.
Simplify standard operating procedures and maximize operator adaptability; all in a compact design for easy storage in confined areas.
Designed for small area cleaning and disinfecting, TruCLEAN 2’s bucket-in-bucket design isolates soiled contaminants from cleaning and disinfecting agents. Clean floors, walls, and ceilings with this truly efficient and lightweight mopping system. Temperature-resistant 36 Liter polypropylene bucket includes embossed graduations for accurate measure and 3-inch twin-wheel casters with one locking caster. Easily accessible brake pedal allows parking on uneven floors.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.