Learn More
Perfex TruCLEAN Deluxe Disinfection
Catalog No. p-200003937 Shop All Perfex Corp. Products
Click to view available options
Color:
Green
Orange
White
This item is not returnable. View return policy
Catalog No. 19-003-134
Supplier: Perfex 303R
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Quickly apply disinfecting agents onto floors, walls, and ceilings. Compact design for simplified use in confined areas.
Designed for quick application of disinfecting agents onto floors, walls, and ceilings. Simply dip the mop in solution, press mop on dosage sieve and apply to surface. Compact design is perfect for use in confined areas, allows easy storage with limited space.
Temperature-resistant 36 Liter polypropylene bucket includes embossed graduations for accurate measure and 3-inch twin-wheel casters with one locking caster. Easily accessible brake pedal allows parking on uneven floors. Stainless steel dosage sieve mounts evenly while precisely positioned apertures release excess disinfectant into 36 Liter or waste containment bucket.
Specifications
Dimensions 19 x 19 x 17 in.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.