Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TrueCut™ Cas9 Protein v2

Catalog No. A36496
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
25 μg
500 μg
Concentration:
1 μg/μL
5 μg/μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
A36496 10 μg 1 μg/μL
A36498 100 μg 5 μg/μL
A36497 25 μg 1 μg/μL
A36499 500 μg 5 μg/μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A36496 Supplier Invitrogen™ Supplier No. A36496
Add to Cart
Edge
Add to Cart

A next-generation CRISPR Cas9 protein engineered to deliver maximum on-target editing efficiency. TrueCut Cas9 Protein v2 is ideal for applications requiring the highest editing efficiency.

Invitrogen TrueCut Cas9 Protein v2 is a next-generation CRISPR Cas9 protein engineered to deliver maximum editing efficiency.

Features include:

  • Consistently high editing efficiency in all tested cell lines including standard, immune, primary, and stem, with up to 2X higher editing efficiency in difficult targets than the competition
  • High quality—manufactured under strict ISO 13485 quality standards
  • Validated protocols for a large number of cell types help you achieve success faster

TrueCut Cas9 Protein v2 is recombinant Streptococcus pyogenes Cas9 (wt) protein, purified from E. coli, for genome editing with CRISPR technology. Cas9 protein forms a very stable ribonucleoprotein (RNP) complex with the guide RNA (gRNA) component of the CRISPR/Cas9 system. Incorporation of nuclear localization signals (NLS) aids delivery to the nucleus, increasing the rate of genomic DNA cleavage.

  • Transfection-ready, using either lipid-mediated transfection reagents or electroporation
  • Eliminates time-consuming cloning steps
  • Available in two concentrations:
    • 1 μg/μL for use in standard editing scenarios
    • 5 μg/μL for optimization of editing conditions in more difficult scenarios such as in primary or embryonic cells, microinjection, or when screening multiple gRNA sequences at a time
    • For custom sizes or concentrations, please inquire

Options for obtaining CRISPR gRNA for use with TrueCut Cas9 Protein v2:

  • Order pre-designed TrueGuide Synthetic gRNAs for high-efficiency knock out in human or mouse
  • Submit your own design for custom synthesis of CRISPR sgRNA
  • Use our free TrueDesign CRISPR Genome Editor software for easy design and ordering of custom gRNA for precise genome editing

Certifications

CoA

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 μg/μL
Components TrueCut Cas9 Protein v2
Grade Molecular Biology
Content And Storage TrueCut Cas9 Protein v2 (1 mg/mL), 10 μg

Store at -5°C to -30°C.
Accession Number NC_002737.2:854751-858857
Expression System E. coli
Gene ID (Entrez) 901176
Gene Symbol Spy_1046
Gene Alias cas5, csn1
Protein Form Recombinant
Protein Tag Untagged
Shipping Condition Dry Ice
Validated Application Genome Editing
Quantity 10 μg
Product Type Cas9 Protein v2
Research Category Genome Editing
Product Line TrueCut
Show More Show Less
What is the molecular weight of TrueCut Cas9 Protein v2?

The molecular weight of TrueCut Cas9 Protein v2 is ~163 kD

What is the difference between TrueCut Cas9 Protein (Prototype) and TrueCutCas9 Protein v2?

Both products are the same CRISPR-Cas9 protein, but the TrueCut Cas9 Protein is manufactured with a new production process and formulated to meet the standards for Ancillary Materials for Cell and Tissue-Based Products.

Is there a specific antibody that you recommend to test the delivery of TrueCut Cas9 Protein v2?

Although we do not have a specific antibody recommendation to test the delivery of TrueCut Cas9 Protein v2, we have tested the following antibodies against an older version of Cas9 (Platinum Cas9) using U2OS-Cas9 expression cell lines (generated using Cas9 lentivirus):
- Cas9 Recombinant Polyclonal Antibody (Cat. No. 711061)
- Cas9 Monoclonal Antibody (Cat.No. MA1-201)
- Cas9 Monoclonal Antibody (Cat. No. MA1-202)

Our TrueCut Cas9 Protein v2 and the Platinum Cas9 have similar domains, and the TrueCut Cas9 Protein v2 has better efficiencies. Therefore, these antibodies should work with TrueCut Cas9 Protein v2 as well.

What is the storage buffer composition for TrueCut Cas9 Protein v2 and TrueCut HiFi Cas9 Protein?

The storage buffer composition for TrueCut Cas9 Protein v2 and TrueCut HiFi Cas9 Protein is as follows:
15 mM Tris-HCl pH 8.0, 150 mM NaCl, 1.0 mM TCEP, 50% glycerol.

Which lipid transfection reagent do you recommend using with TrueCut Cas9 Protein v2 and TrueCut HiFi Cas9 Protein?

We recommend using Lipofectamine CRISPRMAX Transfection Reagent with TrueCut Cas9 Protein v2 or TrueCut HiFi Cas9 Protein.

Do you offer the TrueCut Cas9 Protein v2 at different concentrations?

Yes, we offer TrueCut Cas9 Protein v2 in the following concentrations: 1 µg/µL (Cat. Nos. A36496, A36497) for use in standard editing scenarios, 5 µg/µL (Cat. Nos. A36498, A36499) for optimization of editing conditions in more difficult scenarios such as in primary or embryonic cells, microinjection, or when screening multiple gRNA sequences at a time

What gRNA format do you recommend using with TrueCut Cas9 Protein v2 and TrueCut HiFi Cas9 Protein?

TrueCut Cas9 Protein v2 and TrueCut HiFi Cas9 Protein are designed to work with TrueGuide synthetic gRNAs, but can also be used with in vitro transcribed gRNA from the GeneArt Precision gRNA Synthesis Kit.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.