Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SW™ Sustainability Solutions TrueForm™ TF-95BL Blue 3.5 mil Sustainable Nitrile Exam Gloves
SDP

Catalog No. 19021881
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large

Value and performance hand protection with EcoTek™ sustainable technology.

  • EcoTek™ sustainable technology
  • EnerGel™ Hand Conditioning Technology clinically proven to hydrate and soothe skin
  • Proprietary coating actively manages the skin’s natural pH balance™ to discourage microbial growth that can lead to infection
  • Finger textured surface for wet and dry grip
  • Low Dermatitis Potential (LDP) is an FDA approved claim that gloves were clinically tested and have a reduced potential to cause skin irritation
  • Exceeds 1.5 AQL laboratory standards
TRUSTED_SUSTAINABILITY

Specifications

Accelerator Free No
Applications Healthcare
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 3.5 mil
Finger Thickness (Metric) 0.08 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 2.4 mil
Palm Thickness (Metric) 0.06 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size 2X-Large
Sterility Non-sterile
Thickness (English) 2.4 mil
Thickness (Metric) 0.06 mm
Model N091346
Series TF-95RB
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.