Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TrueMark™ Amplification Control I (1 x 105 copies/μL)

Catalog No. A55699
Encompass_Preferred
Change view
Click to view available options
Quantity:
1000 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55699 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A55699 Supplier Applied Biosystems™ Supplier No. A55699
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The TrueMark Amplification Control I for TrueMark plates contains a linearized multi-target plasmid pool with microbial target sequences used on TrueMark infectious disease research panels.

  • TrueMark Amplification Control I contains a linearized multi-target plasmid pool with microbial target sequences used on TrueMark infectious disease research panels
  • The control has targets relevant to respiratory, urinary, vaginal, and genital microbiota research
  • It can be included as a positive control for panel-specific amplification
  • The higher concentration (5 x 107 copies/) can be used to determine parameters necessary for analytical validation (e.g., limit of detection and Ct cut-off values)
  • The plasmid also contains target sequences for TaqMan Universal Extraction Control Organism (Bacillus atrophaeus) and the human RNase P gene
  • During real-time PCR, the control can be used as a stand-alone sample to verify assay performance and to help with troubleshooting
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 x 105 Copies/μL
Content And Storage Store at -15°C to -25°C.
Detection Method Primer-probe
Format Tube
Product Line TaqMan
Quantity 1000 μL
Shipping Condition Dry ice
Volume 1000 μL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.