Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TrueMark™ Enteric Bacterial Select Panel II Combo Kit

Catalog No. A58076
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A58076 200 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A58076 Supplier Applied Biosystems™ Supplier No. A58076
Add to Cart
Add to Cart

The TrueMark Enteric Bacterial Select Panel II Combo Kit includes the TrueMark Enteric Bacterial Select Panel II and TrueMark Infectious Disease 1‐Step Multiplex Master Mix for simplified ordering.

The TrueMark Enteric Bacterial Select Panel II Combo Kit includes the TrueMark Enteric Bacterial Select Panel II and TrueMark Infectious Disease 1‐Step Multiplex Master Mix for simplified ordering. Together with the TrueMark Enteric Bacterial Select Amplification Control II (offered separately), the kit can be used to quickly evaluate and research up to 200 samples consisting of extracted nucleic acid from stool specimens.

The kit is recommended for use with the QuantStudio 5 Real-Time PCR System (with 96-well, 0.2-mL block). It includes primers and probe sets specific to the following targets:
• Vibrio (V. parahaemolyticus, V. vulnificus, and V. cholerae)
Yersinia enterocolitica
Plesiomonas shigelloides
Aeromonas hydrophila
Bacillus atrophaeus (used as a process control)

Features of the TrueMark Enteric Bacterial Select Panel II Combo Kit include:
• Detects and differentiates targets found in enteric bacterial infections
• Affordable and scalable—add to existing infrastructure to expand research menu while maintaining low operational costs
• Ready-to-use off-the-shelf solution—seamlessly integrate test into existing workflow

The TrueMark Enteric Bacterial Select Panel II Combo Kit includes the following components:
TrueMark Enteric Bacterial Select Panel II—multiplexed assay that contains primer and probe sets
TrueMark Infectious Disease 1‐Step Multiplex Master Mix (No ROX)—ready-to-use PCR mix, including polymerase, dNTPs, salt, and buffer

Use this kit with:
• TrueMark Enteric Bacterial Select Amplification Control II (Cat. No. A58139)—plasmid control containing target-specific templates

TRUSTED_SUSTAINABILITY

Specifications

Detection Method qPCR
Form Liquid
PCR Method Multiplex qPCR, qPCR
Reaction Speed 120 min.
Quantity 200 reactions
For Use With (Application) Pathogen Detection, Bacterial Enteric Diagnostic Testing, Real Time PCR (qPCR), RT-PCR
No. of Reactions 200 Reactions
Reaction Format Single tubes
Sample Type Stool

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.