Learn More
Applied Biosystems™ TrueMark™ STI Select Panel Combo Kit
Description
The Applied Biosystems TrueMark STI Select Panel Combo Kit is a multiplex real‐time PCR solution that contains the assay and master mix needed for the simultaneous detection of nucleic acid from Chlamydia trachomatis (CT), Neisseria gonorrhoeae (NG), Mycoplasma genitalium (MG), and Trichomonas vaginalis (TV) in extracted human samples. Human ribonuclease P (RNase P) is included as endogenous sample collection control.
Features of the TrueMark STI Select Panel Combo Kit include:
• Single test for CT, NG, MG, TV; detect and differentiate between clinically relevant targets found in sexually transmitted infections
• Affordable and scalable—add to existing infrastructure to expand menu while maintaining low operational costs
• Ready-to-use off-the-shelf solution—seamlessly integrate test into the existing workflow
• Research Use Only
The TrueMark STI Select Panel Combo Kit includes the TrueMark STI Select Panel and TrueMark Infectious Disease 1‐Step Multiplex Master Mix for simplified ordering. Together with the TrueMark STI Amplification Control, the kit can be used to quickly evaluate up to 200 samples and is validated for use with RNA extracted from vaginal swabs. The kit is recommended for use with the MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit, the KingFisher Flex Purification System, and the QuantStudio 5 Real-Time PCR System (with 96-well, 0.2-mL block).
The TrueMark STI Select Panel Combo Kit includes the following components:
• TrueMark STI Select Panel—multiplexed assay that contains primer and probe sets specific to CT, NG, MG, TV, and RNase P targets
• TrueMark Infectious Disease 1‐Step Multiplex Master Mix (No ROX)
Use this kit with:
• TrueMark STI Amplification Control (Cat. No. A57009)—plasmid control containing templates specific to CT, NG, MG, TV, and RNase P
Specifications
Specifications
| Detection Method | Primer-probe |
| For Use With (Equipment) | QuantStudio™ 5 Real‐Time PCR System, 96-well, 0.2-mL block |
| Quantity | 200 reactions |
| For Use With (Application) | Pathogen Detection |
| No. of Reactions | 200 Reactions |
Frequently Asked Questions (FAQs)
No, the assay IDs for the individual target assays in the TrueMark STI Select Panel are not provided separately.
Yes, RNase P is the human internal sample control target utilized in the TrueMark, STI Select Panel, Combo Kit and it is one of the multiplexed assays in the kit.
We recommend running the TrueMark STI Select Panel, Combo Kit on the QuantStudio 5 Real-Time PCR instrument, 96-well, 0.2 mL block though other instruments that have the ability to detect Alexa Fluor 647 dye are capable of running the kit. Information about other reagents and consumables that are needed can be found in the user guide ((https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0028493-TrueMarkSTISelectPanel-UG-RUO-EN.pdf).
No, these are single-tube assays and the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0028493-TrueMarkSTISelectPanel-UG-RUO-EN.pdf) provides details on how to setup and create a template to detect and assign the targets to each well.
The TrueMark STI Select Panel, Combo Kit (Cat. No. A57083) contains the TrueMark STI Select Panel (Part No. A52691) and the TrueMark Infectious Disease 1-Step Multiplex Master Mix (No ROX) (Part No. A57052). Other required materials are standard qPCR consumables and instruments and they can be found in the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0028493-TrueMarkSTISelectPanel-UG-RUO-EN.pdf).
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.