Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRUSS Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220741
Description
TRUSS Polyclonal specifically detects TRUSS in Human samples. It is validated for Western Blot.Specifications
TRUSS | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
C20orf188, dJ756N5.2, DKFZp586C1223, DKFZP727M231, Protein TAP1, Protein TRUSS, short transient receptor potential channel 4 associated protein, short transient receptor potential channel 4-associated protein, TNF-receptor ubiquitous scaffolding/signaling protein, transient receptor potential cation channel, subfamily C, member 4 associatedprotein, Trp4-associated protein, Trpc4-associated protein, TRRP4APchromosome 20 open reading frame 188, TRUSS, tumor necrosis factor receptor-associated ubiquitous scaffolding and signalingprotein | |
Rabbit | |
91 kDa | |
0.1 mL | |
Primary | |
Human. Species reactivity based on sequence identity: Mouse (100%), Chicken (83%), Rat (100%), Xenopus laevis (83%), Bovine (100%), Xenopus tropicalis (82%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
Q8TEL6 | |
TRPC4AP | |
Recombinant protein encompassing a sequence within the N-terminus region of human TRUSS. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
26133 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction