Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRUSS Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | TRUSS |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
TRUSS Polyclonal specifically detects TRUSS in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
TRUSS | |
Polyclonal | |
Rabbit | |
Human | |
26133 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ESSFRFWQARAVESFLRGTTSYADQMFLLKRGLLEHILYCIVDSECKSRDVLQSYFDLLGELMKFNVDAFKRFNKYINTDAKFQVFLKQINS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
C20orf188, dJ756N5.2, DKFZp586C1223, DKFZP727M231, Protein TAP1, Protein TRUSS, short transient receptor potential channel 4 associated protein, short transient receptor potential channel 4-associated protein, TNF-receptor ubiquitous scaffolding/signaling protein, transient receptor potential cation channel, subfamily C, member 4 associatedprotein, Trp4-associated protein, Trpc4-associated protein, TRRP4APchromosome 20 open reading frame 188, TRUSS, tumor necrosis factor receptor-associated ubiquitous scaffolding and signalingprotein | |
TRPC4AP | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title