Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TrxR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111192
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111192 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111192 Supplier Invitrogen™ Supplier No. PA5111192
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GFDQDMANKI GEHMEEHGIK FIRQFVPIKV EQIEAGTPGR LRVVAQSTNS EEIIEGEYNT VML Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 86%.

This gene encodes a member of the family of pyridine nucleotide oxidoreductases. This protein reduces thioredoxins as well as other substrates, and plays a role in selenium metabolism and protection against oxidative stress. The functional enzyme is thought to be a homodimer which uses FAD as a cofactor. Each subunit contains a selenocysteine (Sec) residue which is required for catalytic activity. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenocysteine-containing genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Alternative splicing results in several transcript variants encoding the same or different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TrxR1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TXNRD1
Gene Accession No. Q16881
Gene Alias gene associated with retinoic and IFN-induced mortality 12 protein; gene associated with retinoic and interferon-induced mortality 12 protein; GRIM12; GRIM-12; KDRF; KM-102-derived reductase-like factor; MGC9145; NADPH-dependent thioredoxin reductase; oxidoreductase; selenoprotein oxidoreductase; testis tissue sperm-binding protein Li 46a; thioredoxin reductase 1; thioredoxin reductase 1, cytoplasmic; thioredoxin reductase GRIM-12; thioredoxin reductase TR1; TR; TR alpha; TR1; Trxr1; TXNR; Txnrd1
Gene Symbols TXNRD1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TrxR1. Recombinant protein control fragment (Product #RP-104781).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7296
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.