Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TrxR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116094
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116094 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116094 Supplier Invitrogen™ Supplier No. PA5116094
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TXNRD1.

This gene encodes a member of the family of pyridine nucleotide oxidoreductases. This protein reduces thioredoxins as well as other substrates, and plays a role in selenium metabolism and protection against oxidative stress. The functional enzyme is thought to be a homodimer which uses FAD as a cofactor. Each subunit contains a selenocysteine (Sec) residue which is required for catalytic activity. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenocysteine-containing genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Alternative splicing results in several transcript variants encoding the same or different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TrxR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene TXNRD1
Gene Accession No. O89049, Q16881, Q9JMH6
Gene Alias gene associated with retinoic and IFN-induced mortality 12 protein; gene associated with retinoic and interferon-induced mortality 12 protein; GRIM12; GRIM-12; KDRF; KM-102-derived reductase-like factor; MGC9145; NADPH-dependent thioredoxin reductase; oxidoreductase; selenoprotein oxidoreductase; testis tissue sperm-binding protein Li 46a; thioredoxin reductase 1; thioredoxin reductase 1, cytoplasmic; thioredoxin reductase GRIM-12; thioredoxin reductase TR1; TR; TR alpha; TR1; Trxr1; TXNR; Txnrd1
Gene Symbols TXNRD1
Host Species Rabbit
Immunogen A synthesized peptide derived from human TXNRD1(Accession Q16881), corresponding to amino acid residues Y302-D323.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50493, 58819, 7296
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.