Trypan Blue, dye content >60%

Manufacturer: Alfa Aesar A1860014

Quantity 25g

 View more versions of this product

Catalog No. AAA1860014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 72-57-1
    Molecular Formula C34H24N6Na4O14S4
    Formula Weight 960.82
    MDL Number MFCD00003969
    Merck Index 14,9792
    Chemical Name or Material Trypan Blue, dye content >60%

    Specifications

    Formula Weight 960.82
    Quantity 25g
    MDL Number MFCD00003969
    Merck Index 14,9792
    Health Hazard 1 Danger
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H350
    May cause cancer.
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or c
    RTECS Number QJ6475000
    Molecular Formula C34H24N6Na4O14S4
    CAS 72-57-1
    Chemical Name or Material Trypan Blue, dye content >60%

    Description

    Applications
    Trypan blue is commonly used in microscopy (for cell counting) and in laboratory mice for assessment of tissue viability. Used in dye exclusion studies in cell culture. A viability dye for quantifying live versus dead cells with permeable membranes, blue. It is also used to identify islet cell cluster cell viability from caprine pancreatic samples. It is also useful to observe hyphea of fungi and Stramenopiles. Trypan blue solution may be used in trypan blue based cytotoxitiy and proliferation assays.

    Solubility
    Soluble in water (10 mg/ml at 25°C), ethanol (0.6 mg/ml), and EGME (20 mg/ml).

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    GHS H Statement
    H350
    May cause cancer.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or c

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit