Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Trypsin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119849
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119849 Supplier Invitrogen™ Supplier No. PA5119849
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Human small intestine tissue lysates, rat pancreas tissue, human pancreas tissue, mouse pancreas tissue, SH-SY5Y. Subcellular Location: Extracellular space.

This gene belongs to the trypsin family of serine proteases and encodes anionic trypsinogen. It is part of a cluster of trypsinogen genes that are located within the T cell receptor beta locus. Enzymes of this family cleave peptide bonds that follow lysine or arginine residues. This protein is found at high levels in pancreatic juice and its upregulation is a characteristic feature of pancreatitis. This protein has also been found to activate pro-urokinase in ovarian tumors, suggesting a function in tumor invasion. In addition, this enzyme is able to cleave across the type II collagen triple helix in rheumatoid arthritis synovitis tissue, potentially participating in the degradation of type II collagen-rich cartilage matrix. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Trypsin 2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene PRSS2
Gene Accession No. P00763, P07146, P07478
Gene Alias anionic trypsin; anionic trypsin II; Anionic trypsin-2; anionic trypsinogen; EC 3.4.21; EC 3.4.21.4; MGC111183; MGC120174; MGC120175; pancreatic anionic trypsinogen; pancreatic trypsin 2; pancreatic trypsin II; pretrypsinogen II; protease serine 2 preproprotein; protease, serine 2; protease, serine, 2; protease, serine, 2 (trypsin 2); PRSS2; Ptryss2; Serine protease 2; Ta; Tesp4; Try2; TRY8; TRYP; TRYP2; TRYP8; trypsin 2; trypsin II; trypsin Trypsin II; Trypsin-2; trypsinogen 2; trypsinogen 20; trypsinogen anionic precursor
Gene Symbols PRSS2
Host Species Rabbit
Immunogen Recombinant protein within human PRSS2 aa 1-120.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22072, 25052, 5645
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.