Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trypsin 2/PRSS2 Antibody (12H7), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Trypsin 2/PRSS2 |
---|---|
Clone | 12H7 |
Dilution | ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | ELISA, Immunocytochemistry |
Classification | Monoclonal |
Description
Trypsin 2/PRSS2 Monoclonal antibody specifically detects Trypsin 2/PRSS2 in Human samples. It is validated for ELISA, ImmunocytochemistrySpecifications
Trypsin 2/PRSS2 | |
ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Anionic trypsinogen, EC 3.4.21, EC 3.4.21.4, MGC111183, MGC120174, MGC120175, protease serine 2 preproprotein, protease, serine, 2 (trypsin 2), protease, serine, 2, preproprotein, Serine protease 2, TRY2TRY8, TRYP2, trypsin 2, Trypsin II, trypsin-2, trypsinogen 2 | |
A synthesized peptide derived from Human Trypsin 2/PRSS2 [UniProt P07478] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
12H7 | |
ELISA, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Cell Biology | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
5645 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title