Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Trypsin 2/PRSS2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3586SP
This item is not returnable.
View return policy
Description
Trypsin 2/PRSS2 Monoclonal specifically detects Trypsin 2/PRSS2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Trypsin 2/PRSS2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
P07478 | |
PRSS2 | |
Mouse myeloma cell line NS0-derived recombinant human Trypsin 2/PRSS2 Ala16-Ser247 Accession # P07478 | |
25 μg | |
Primary | |
Detects human Trypsin 2/PRSS2 in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) Trypsin 1 or rhTrypsin 3. Recognizes both the pro and mature forms of rhTrypsin 2. | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
410322 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Anionic trypsinogen, EC 3.4.21, EC 3.4.21.4, MGC111183, MGC120174, MGC120175, protease serine 2 preproprotein, protease, serine, 2 (trypsin 2), protease, serine, 2, preproprotein, PRSS2, Serine protease 2, Tesp4, TRY2TRY8, TRY8, TRYP2, trypsin 2, Trypsin II, trypsin-2, trypsinogen 2, Trypsinogen 20 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5645 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction