Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trypsin 3/PRSS3 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299895MFV500
Description
Trypsin 3/PRSS3 Polyclonal antibody specifically detects Trypsin 3/PRSS3 in Human samples. It is validated for ELISASpecifications
Trypsin 3/PRSS3 | |
Polyclonal | |
ELISA | |
Brain trypsinogen, EC 3.4.21, EC 3.4.21.4, Mesotrypsinogen, pancreatic trypsinogen III, protease, serine, 3, protease, serine, 3 (mesotrypsin), protease, serine, 4 (trypsin 4, brain), PRSS4, Serine protease 3, Serine protease 4, T9, TRY3MTG, TRY4mesotrypsin, trypsin 3, Trypsin III, Trypsin IV, trypsin-3, trypsinogen 4, trypsinogen 5, trypsinogen IV | |
Produced in rabbits immunized with purified, recombinant Human Trypsin 3/PRSS3 (Accession#: P35030-3; Met 1-Ser 247) | |
0.1 mL | |
Cancer, Proteases & Other Enzymes | |
5646 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction