Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trypsin Inhibitor Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178088H
Description
Trypsin Inhibitor Polyclonal antibody specifically detects Trypsin Inhibitor in Plant samples. It is validated for Western Blot, ELISASpecifications
Trypsin Inhibitor | |
Polyclonal | |
Western Blot, ELISA | |
KTI3, Kunitz trypsin inhibitor | |
Trypsin Inhibitor [Lima Bean] (Uniprot: P01070) | |
0.1 mL | |
Primary | |
Plant | |
IgG |
Western Blot, ELISA | |
HRP | |
PBS | |
Rabbit | |
Multi-step | |
RUO | |
547831 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction