Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trypsin Monoclonal Antibody (TRYP11-224.3), QED Bioscience™
SDP

Catalog No. 502277521
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7521 1 mg
50-227-7522 100 μg
50-227-7523 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 50-227-7521 Supplier Leinco Technologies Inc Supplier No. 134031000UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Trypsin Monoclonal antibody specifically detects Trypsin in Human samples. It is validated for ELISA

Specifications

Antigen Trypsin
Applications ELISA
Classification Monoclonal
Clone TRYP11-224.3
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PRSS1
Gene Accession No. P07477
Gene Alias Alpha-trypsin chain 1; Alpha-trypsin chain 2; Anionic trypsin I; anionic trypsin-1; Anionic trypsinogen; beta-trypsin; Cationic trypsinogen; digestive zymogen; MGC120175; MGC149362; nonfunctional trypsin 1; pancreatic trypsin 1; pretrypsinogen I; Protease Serine 1; protease serine 2 preproprotein; protease, serine 1; protease, serine 1 (trypsin 1); protease, serine 2; protease, serine, 1; protease, serine, 1 (trypsin 1); protease, serine, 2; protease, serine, 2 (trypsin 2); PRSS1; PRSS2; PTRYI; RATPTRYI; Serine protease 1; Serine protease 2; TCR V beta 4.1; TRP1; Try1; Try-1; TRY2; TRY4; TRY8; Trygn16; TRYP1; TRYP2; TRYP8; trypsin 1; Trypsin I; Trypsin II; trypsin-1; Trypsin-2; trypsinogen; trypsinogen 1; trypsinogen 16; trypsinogen 2; trypsinogen A
Host Species Mouse
Immunogen Purified human pancreatic trypsin.
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5644
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.