Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Trypsin Pan Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546939
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546939 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546939 Supplier Invitrogen™ Supplier No. PA546939
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Trypsin is a 24 kDa enzyme of the serine proteinase family. It is produced in the pancreas as an inactive precursor, trypsinogen, but the active enzyme is located in the gastrointestinal tract where it degrades proteins to large peptides. Trypsin prefers lysine and arginine residues. One of the substrates for trypsin is chymotrypsinogen which is cleaved to produce active chymotrypsin. High levels of immunoreactive trypsin in the bloodstream can indicate pancreatic malfunction and this indicator is used to screene for and diagnose Cystic Fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Trypsin Pan
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene PRSS3
Gene Accession No. P07477, P07478, P35030
Gene Alias Alpha-trypsin chain 1; Alpha-trypsin chain 2; Anionic trypsinogen; beta-trypsin; Brain trypsinogen; cationic trypsinogen; digestive zymogen; mesotrypsin; Mesotrypsinogen; MTG; nonfunctional trypsin 1; pancreatic trypsinogen III; protease serine 2 preproprotein; protease, serine 1; protease, serine 2; protease, serine 3; protease, serine, 2 (trypsin 2); protease, serine, 4 (trypsin 4, brain); PRSS1; PRSS2; PRSS3; PRSS4; Serine protease 1; Serine protease 2; Serine protease 3; Serine protease 4; T9; TCR V beta 4.1; TRP1; TRY1; TRY2; TRY3; TRY4; TRY8; TRYP1; TRYP2; Trypsin I; Trypsin II; trypsin III; Trypsin IV; trypsin-1; Trypsin-2; trypsin-3; trypsinogen 1; trypsinogen 2; trypsinogen 4; trypsinogen 5; trypsinogen A; trypsinogen IV
Gene Symbols PRSS1, PRSS2, PRSS3
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Trypsin 2 Ala16-Ser247.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5644, 5645, 5646
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.