Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trypsin Polyclonal Antibody, Invitrogen™

Catalog No. PIPA588532
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588532 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588532 Supplier Thermo Scientific Supplier No. PA588532

Rabbit Polyclonal Antibody

Immunogen sequence: IVGGYNCEEN SVPYQVSLNS GYHFCGGSLI NEQWVVSAGH CYKSRIQVRL GEHNIEVLEG NEQFINAAKI IRHPQYDRKT LNNDIMLIKL SSRAVINARV STISLPTAPP ATGTKCLISG WGNTASSGAD YPDELQCLDA PVLSQAKCEA SYPGKITSNM FCVGFLEGGK DSCQGDSGGP VVCNGQLQGV VSWGDGCAQK NKPGVYTKVY NYVKWIKNTI AANS; Positive Samples: HT-29, Mouse spleen, Mouse kidney; Cellular Location: extracellular space

Trypsin is a 24 kDa enzyme of the serine proteinase family. It is produced in the pancreas as an inactive precursor, trypsinogen, but the active enzyme is located in the gastrointestinal tract where it degrades proteins to large peptides. Trypsin prefers lysine and arginine residues. One of the substrates for trypsin is chymotrypsinogen which is cleaved to produce active chymotrypsin. High levels of immunoreactive trypsin in the bloodstream can indicate pancreatic malfunction and this indicator is used to screene for and diagnose Cystic Fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Trypsin
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 4.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRSS1
Gene Accession No. P07477
Gene Symbols Prss1
Host Species Rabbit
Immunogen Recombit fusion protein containing a sequence corresponding to amino acids 24-247 of human PRSS1
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114228, 5644
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.