Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Trypsin Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. FEPA5145273
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145273 200 μL
FEPA5145274 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145273 Supplier Invitrogen™ Supplier No. PA5145273
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Trypsin is a 24 kDa enzyme of the serine proteinase family. It is produced in the pancreas as an inactive precursor, trypsinogen, but the active enzyme is located in the gastrointestinal tract where it degrades proteins to large peptides. Trypsin prefers lysine and arginine residues. One of the substrates for trypsin is chymotrypsinogen which is cleaved to produce active chymotrypsin. High levels of immunoreactive trypsin in the bloodstream can indicate pancreatic malfunction and this indicator is used to screene for and diagnose Cystic Fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Trypsin
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene PRSS1
Gene Accession No. P00762, P07477
Gene Alias Alpha-trypsin chain 1; Alpha-trypsin chain 2; Anionic trypsin I; anionic trypsin-1; Anionic trypsinogen; beta-trypsin; Cationic trypsinogen; digestive zymogen; MGC120175; MGC149362; nonfunctional trypsin 1; pancreatic trypsin 1; pretrypsinogen I; Protease Serine 1; protease serine 2 preproprotein; protease, serine 1; protease, serine 1 (trypsin 1); protease, serine 2; protease, serine, 1; protease, serine, 1 (trypsin 1); protease, serine, 2; protease, serine, 2 (trypsin 2); PRSS1; PRSS2; PTRYI; RATPTRYI; Serine protease 1; Serine protease 2; TCR V beta 4.1; TRP1; Try1; Try-1; TRY2; TRY4; TRY8; Trygn16; TRYP1; TRYP2; TRYP8; trypsin 1; Trypsin I; Trypsin II; trypsin-1; Trypsin-2; trypsinogen; trypsinogen 1; trypsinogen 16; trypsinogen 2; trypsinogen A
Gene Symbols PRSS1
Host Species Rabbit
Immunogen Recombinant Human PRSS1 protein, Ser51-Val228 (Accession #P07477).
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114228, 24691, 5644
Target Species Human, Mouse, Rat, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.