Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Trypsin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145472
Description
Trypsin is a 24 kDa enzyme of the serine proteinase family. It is produced in the pancreas as an inactive precursor, trypsinogen, but the active enzyme is located in the gastrointestinal tract where it degrades proteins to large peptides. Trypsin prefers lysine and arginine residues. One of the substrates for trypsin is chymotrypsinogen which is cleaved to produce active chymotrypsin. High levels of immunoreactive trypsin in the bloodstream can indicate pancreatic malfunction and this indicator is used to screene for and diagnose Cystic Fibrosis.
Specifications
Trypsin | |
Polyclonal | |
Unconjugated | |
PRSS1 | |
Alpha-trypsin chain 1; Alpha-trypsin chain 2; Anionic trypsin I; anionic trypsin-1; Anionic trypsinogen; beta-trypsin; Cationic trypsinogen; digestive zymogen; MGC120175; MGC149362; nonfunctional trypsin 1; pancreatic trypsin 1; pretrypsinogen I; Protease Serine 1; protease serine 2 preproprotein; protease, serine 1; protease, serine 1 (trypsin 1); protease, serine 2; protease, serine, 1; protease, serine, 1 (trypsin 1); protease, serine, 2; protease, serine, 2 (trypsin 2); PRSS1; PRSS2; PTRYI; RATPTRYI; Serine protease 1; Serine protease 2; TCR V beta 4.1; TRP1; Try1; Try-1; TRY2; TRY4; TRY8; Trygn16; TRYP1; TRYP2; TRYP8; trypsin 1; Trypsin I; Trypsin II; trypsin-1; Trypsin-2; trypsinogen; trypsinogen 1; trypsinogen 16; trypsinogen 2; trypsinogen A | |
Rabbit | |
Protein A | |
RUO | |
114228, 24691, 5644 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
P00762, P07477 | |
PRSS1 | |
Recombinant Human PRSS1 protein, Ile24-Ser247 (Accession #P07477). | |
20 μL | |
Primary | |
Human, Mouse, Rat, Pig | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction