Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trypsinogen Polyclonal Antibody, Biotin, Rockland™
SDP

Catalog No. 501991202
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1202 100 μg
50-199-1203 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1202 Supplier Rockland Immunochemicals Supplier No. 20046800100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Trypsinogen antibody is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Biotin, anti-Rabbit Serum as well as purified and partially purified Trypsinogen [Bovine Pancreas]. Cross reactivity against Trypsinogen from other sources may occur but has not been specifically determined.

Trypsin is a 24 kDa enzyme of the serine proteinase family. It is produced in the pancreas as an inactive precursor, trypsinogen, but the active enzyme is located in the gastrointestinal tract where it degrades proteins to large peptides. Trypsin prefers lysine and arginine residues. One of the substrates for trypsin is chymotrypsinogen which is cleaved to produce active chymotrypsin. High levels of immunoreactive trypsin in the bloodstream can indicate pancreatic malfunction and this indicator is used to screene for and diagnose Cystic Fibrosis.

Specifications

Antigen Trypsinogen
Applications Dot Blot, ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% sodium azide; pH 7.2
Gene PRSS2
Gene Alias Alpha-trypsin chain 1; Alpha-trypsin chain 2; Anionic trypsin I; Anionic trypsin II; Anionic trypsin-1; Anionic trypsin-2; Anionic trypsinogen; Beta-trypsin; Cationic trypsinogen; digestive zymogen; MGC120175; MGC149362; nonfunctional trypsin 1; pancreatic trypsin 1; pancreatic trypsin 2; pancreatic trypsin II; Pretrypsinogen I; Pretrypsinogen II; Protease Serine 1; protease serine 2 preproprotein; protease, serine 1; protease, serine 1 (trypsin 1); protease, serine 2; protease, serine, 1; protease, serine, 1 (trypsin 1); protease, serine, 2; protease, serine, 2 (trypsin 2); Prss1; PRSS2; PTRYI; Ptryss2; RATPTRYI; Serine protease 1; Serine protease 2; Ta; TCR V beta 4.1; Tesp4; TRP1; TRY1; Try-1; TRY2; TRY4; TRY8; Trygn16; TRYP; TRYP1; TRYP2; TRYP8; trypsin 1; trypsin 2; Trypsin I; Trypsin II; Trypsin-1; Trypsin-2; trypsinogen 1; trypsinogen 16; trypsinogen 2; trypsinogen 20; trypsinogen A
Gene Symbols PRSS1, PRSS2
Host Species Rabbit
Immunogen Trypsinogen [Bovine Pancreas]
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282603, 615026
Target Species Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.