Alfa Aesar™ Tryptamine, 98+%

Manufacturer: Alfa Aesar™ A1111618

Quantity 50g

 View more versions of this product

Catalog No. AAA1111618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 61-54-1
    Molecular Formula C10H12N2
    Formula Weight 160.22
    MDL Number MFCD00005661
    Beilstein 125513
    Merck Index 14,9796
    Synonym 3-(2-Aminoethyl)indole; 2-(3-Indolyl)ethylamine
    Chemical Name or Material Tryptamine

    Specifications

    Formula Weight 160.22
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2°C to 8°C
    CAS 61-54-1
    Merck Index 14,9796
    Synonym 3-(2-Aminoethyl)indole; 2-(3-Indolyl)ethylamine
    Health Hazard 1 Warning
    Boiling Point 137°C (0.15mmHg)
    Molecular Formula C10H12N2
    Beilstein 125513
    Melting Point 111° to 116°C
    Flash Point 185°C (365°F)
    RTECS Number NL4020000
    MDL Number MFCD00005661
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 50g
    Chemical Name or Material Tryptamine
    Assay ≥98%

    Description

    Applications
    Tryptamine is commonly used in the preparation of biologically active compounds such as neurotransmitters and psychedelics, multi-layered analysis, used to differentiate and identify biogenic amines1 • in the synthesis of indole ring-terminated thiol, ω-mercaptooctyltryptamide.

    Solubility
    Soluble in DMSO, Methanol. Insoluble in water.

    Notes
    Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit